Learn More
SCYL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553267
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: AFPEDFCRHK VLPQLLTAFE FGNAGAVVLT PLFKVGKFLS AEEYQQKIIP VVVKMFSSTD RAMRIRLLQQ MEQFIQYLDE PTVNTQIFPH VVHGFLDTNP AIREQTVKSM LLLAPKLNEA NLNVELMK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
SCYL1 Polyclonal antibody specifically detects SCYL1 in Human samples. It is validated for Immunocytochemistry.Spezifikation
SCYL1 | |
Polyclonal | |
Unconjugated | |
Scyl1 | |
105 kDa kinase-like protein; 2810011O19Rik; C85140; Coated vesicle-associated kinase of 90 kDa; CVAK90; GKLP; HT019; likely ortholog of mouse N-terminal kinase-like protein; mdf; mfd; Mitosis-associated kinase-like protein NTKL; NKTL; N-terminal kinase-like protein; NTKL; P105; SCAR21; SCY1 like pseudokinase 1; SCY1-like 1 (S. cerevisiae); SCY1-like 1 splice variant 13; SCY1-like 1 splice variant 15; SCY1-like protein 1; SCY1-like, kinase-like 1; Scyl1; TAPK; TEIF; Telomerase regulation-associated protein; Telomerase transcriptional element-interacting factor; telomerase transcriptional elements-interacting factor; teratoma-associated tyrosine kinase; TRAP; ubiquitous protein kinase-like (105 kDa) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96KG9 | |
Scyl1 | |
Recombinant protein corresponding to Human SCYL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only