Learn More
Invitrogen™ Human Sclerostin (SOST) ELISA Kit
Beschreibung
The Human Sclerostin (SOST) ELISA quantitates Hu Sclerostin human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Sclerostin. Principle of the method The Human Sclerostin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Spezifikation
Spezifikation
| Zugriffsnummer | Q9BQB4 |
| Assay-Bereich | 40,96 pg/ml bis 10.000 pg/ml |
| Assay-Empfindlichkeit: | 15 pg/mL |
| Kalibrierbereich | 15 pg/mL bis 10000 pg/mL |
| Assay | Festphasen-Sandwich-ELISA |
| Konjugat | Biotin |
| Produkttyp | ELISA-Kit |
| Proteinfamilie | Weitere Proteine |
| Probentyp | Plasma, Serum, Überstand |
| Spezifität | Dieses ELISA-Antikörperpaar erkennt humanes SOST. Weitere Spezies sind noch nicht bestimmt. |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.