missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12569-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides.Spezifikation
SCFD1 | |
Polyclonal | |
Unconjugated | |
SCFD1 | |
C14orf163, KIAA0917, RA410, SCFD1, SLY1, SLY1 homolog, Sly1p, STXBP1L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23256, 54350, 76983 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62991, Q8BRF7, Q8WVM8 | |
Scfd1 | |
SCFD1 Fusion Protein Ag3234 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |