missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SART1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22675-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody recognizes the endogenous 800-amino acid SART1, which migrates with an apparent molecular mass of 110 kDa in SDS-PAGE.
This gene encodes two proteins, the SART1(800) protein expressed in the nucleus of the majority of proliferating cells, and the SART1(259) protein expressed in the cytosol of epithelial cancers. The SART1(259) protein is translated by the mechanism of -1 frameshifting during posttranscriptional regulation; its full-length sequence is not published yet. The two encoded proteins are thought to be involved in the regulation of proliferation. Both proteins have tumor-rejection antigens. The SART1(259) protein possesses tumor epitopes capable of inducing HLA-A2402-restricted cytotoxic T lymphocytes in cancer patients. This SART1(259) antigen may be useful in specific immunotherapy for cancer patients and may serve as a paradigmatic tool for the diagnosis and treatment of patients with atopy. The SART1(259) protein is found to be essential for the recruitment of the tri-snRNP to the pre-spliceosome in the spliceosome assembly pathway.Spezifikation
| SART1 | |
| Polyclonal | |
| Unconjugated | |
| SART1 | |
| Ara1, Hom s 1, HOMS1, hSART 1, hSnu66, SART 1, SART1, SART1259, SNRNP110, Snu66, SNU66 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20227, 9092 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43290, Q9Z315 | |
| SART1 | |
| SART1 Fusion Protein Ag18478 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur