missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 ORF8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
240.00 CHF - 484.00 CHF
Spezifikation
| Antigen | ORF8 |
|---|---|
| Klon | 1041422 |
| Verdünnung | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Anwendungen | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18741897
![]() |
R&D Systems
MAB10820-SP |
25 μg |
240.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18743527
![]() |
R&D Systems
MAB10820-100 |
100 μg |
484.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
ORF8 Monoclonal antibody specifically detects ORF8 in SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, ImmunocytochemistrySpezifikation
| ORF8 | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| E. coli-derived human SARS-CoV-2 ORF8, Met16-His121, Accession # YP_009724369 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1041422 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| SARS-CoV-2 | |
| YP_009724369 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts