missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-COV-2 ORF1a Polyprotein (aa1564-1878), His Tag Recombinant Protein

E. coli Recombinant Protein

Marke:  Invitrogen™ RP87701

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 17193773

  • 428.00 CHF / 100 Mikrogramm
Verfügbar ab: 21-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Severe acute respiratory syndrome (SARS) is a viral respiratory illness caused by a coronavirus, called SARS-associated coronavirus (SARS-CoV). Human coronaviruses (HCoVs) were previously only associated with mild diseases. The SARS-CoV genome contains five major open reading frames (ORFs) that encode the replicase polyprotein; the spike (S), envelope (E), and membrane (M) glycoproteins; and the nucleocapsid protein (N).
  • Purity: Value and Method: > 90% by SDS-PAGE
  • Endotoxin Concentration: < 1 EU/μg of the protein by LAL method
  • Protein Length/ AA Sequence: Glu1564-Lys1878 of 2019-ncov papain-like protease fused with an initial Met, a 6×His tag at the N-terminus.
  • Molecular Weight: 36.6 kDa
  • Animal Origin Free: Yes
Spezifikation

Spezifikation

Control, ELISA, Western Blot
36.6 kDa
100 μg
-80°C, Avoid Freeze/Thaw Cycles
Polyprotein orf1a; PP1a; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS-CoV-2
Unconjugated
Liquid
20 mM tris with 10 mM beta mercaptoethanol, 20% glycerol and no preservative; pH 7.5
SARS-COV-2 ORF1a Polyprotein (aa1564-1878), His Tag
E. coli
RUO
Virus
Recombinant
>90%
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote