missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP130 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-20259
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SAP130 Polyclonal specifically detects SAP130 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| SAP130 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| KIAA0017splicing factor 3b, subunit 3, 130kD, Pre-mRNA-splicing factor SF3b 130 kDa subunit, RSE1, SAP130SAP 130, SF3B130, SF3b130pre-mRNA splicing factor SF3b, 130 kDa subunit, Spliceosome-associated protein 130, splicing factor 3B subunit 3, splicing factor 3b, subunit 3, 130kDa, STAF130 | |
| Rabbit | |
| 136 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (100%), Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q15393 | |
| SF3B3 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human SAP130. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 23450 | |
| Human, Mouse | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur