Learn More
Cole-Parmer Sanitary Clamp Fittings, Straight Hose Barb Adapter, 316 Stainless Steel
Artikelnummer. p-200105211 Alle ansehen Cole-Parmer Produkte
Click to view available options
Durchmesser (metrisch):
1-1/2 in.
1/2 in.
1/4 in.
1/8 in.
1 in.
2 in.
3/4 in.
3/8 in.
Packungsgröße:
1 Stück
Artikelnummer. 30054121
Marke: Cole-Parmer 5011809
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
Ideal for general-purpose, food, beverage, and pharmaceutical applications. Free technical application support available to assist with tubing selection. 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases. Hose barb connection helps easily transition to tubing
Spezifikation
Zertifizierung/Konformität 3 A;FDA
Durchmesser (metrisch) 3/8 in.
Auslassanschluss Sanitary x Hose Barb
Form Straight
Enthält 3/4 in. Sanitary Clamp Size