Learn More
Cole-Parmer Sanitary Clamp Fittings, Straight Hose Barb Adapter, 316 Stainless Steel
Alle ansehen Cole-Parmer Produkte
Click to view available options
Durchmesser (metrisch):
1-1/2 in.
1/2 in.
1/4 in.
1/8 in.
1 in.
2 in.
3/4 in.
3/8 in.
Packungsgröße:
1 Stück
Beschreibung
Ideal for general-purpose, food, beverage, and pharmaceutical applications. Free technical application support available to assist with tubing selection. 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases. Hose barb connection helps easily transition to tubing
Spezifikation
Spezifikation
| Zertifizierung/Konformität | 3 A;FDA |
| Durchmesser (metrisch) | 3/8 in. |
| Auslassanschluss | Sanitary x Hose Barb |
| Form | Straight |
| Enthält | 3/4 in. Sanitary Clamp Size |