missing translation for 'onlineSavingsMsg'
Learn More

Sancyclinhydrochlorid, TRC

Product Code. 30415900
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30415900 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30415900 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCS11150050MG
 Eingeschränkter Artikel
Verfügbar ab: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6625-20-3
Summenformel C21 H22 N2 O7 . Cl H
Molekulargewicht (g/mol) 450.87
Synonym 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, monohydrochloride, (4S,4aS,5aR,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, monohydrochloride (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5aα,12aα)]-, 6-Demethyl-6-deoxytetracycline hydrochloride, NSC 51812, Sancycline hydrochloride
IUPAC-Name (4S,4aS,5aR,12aS)-4-(Dimethylamino)-3,10,12,12a-Tetrahydroxy-1,11-Dioxo-4a,5,5a,6-tetrahydro-4H-tetracen-2-Carboxamid; Hydrochlorid
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4cccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N

Specifications

Chemischer Name oder Material Sancycline Hydrochloride
Menge 50 mg
InChI-Formel InChI=1S/C21H22N2O7.ClH/c1-23(2)15-10-7-9-6-8-4-3-5-11(24)12(8)16(25)13(9)18(27)21(10,30)19(28)14(17(15)26)20(22)29;/h3-5,9-10,15,24,26-27,30H,6-7H2,1-2H3,(H2,22,29);1H/t9-,10-,15-,21-;/m0./s1
Formelmasse 450.1194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.