missing translation for 'onlineSavingsMsg'
Learn More

(S)-Quizalofop Ethyl, TRC

Product Code. 30415710
Change view
Click to view available options
Menge:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
30415710 250 mg
30408986 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415710 Supplier Toronto Research Chemicals Supplier No. TRCQ164650250MG
 Eingeschränkter Artikel
Verfügbar ab: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100646-52-4
Summenformel C19H17ClN2O4
Molekulargewicht (g/mol) 372.8
Synonym 2-[4-[(6-Chloro-2-quinoxalinyl)oxy]phenoxy]-Ethyl Ester-(S)-Propanoic Acid, (-)-Quizalofop Ethyl
IUPAC-Name Ethyl(2S)-2-[4-(6-Chlorquinoxalin-2-yl)Oxyphenoxy]propanoat
SMILES CCOC(=O)[C@H](C)Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1

Specifications

Chemischer Name oder Material (S)-Quizalofop Ethyl
Menge 250 mg
InChI-Formel InChI=1S/C19H17ClN2O4/c1-3-24-19(23)12(2)25-14-5-7-15(8-6-14)26-18-11-21-17-10-13(20)4-9-16(17)22-18/h4-12H,3H2,1-2H3/t12-/m0/s1
Formelmasse 372.0877
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.