missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ S 2101
Description
S 2101 is a LSD1 inhibitor (IC50 = 990 nM; Ki = 610 nM). Exhibits selectivity for LSD1 over MAO-B and MAO-A (Ki values are 17 and 110 μM, respectively).
Specifications
Specifications
| Chemischer Name oder Material | (1R,2S)-rel-2-[3,5-Difluoro-2-(phenylmethoxy)phenyl]cycloprpanamine hydrochloride |
| CAS | 1239262-36-2 |
| Menge | 50 mg |
| Produktlinie | Tocris |
| Ziel | Histone Demethylase Inhibitors |
| Summenformel | C16H15F2NO.HCl |
| Formelmasse | 311.75 |
| Reinheit | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?