missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXFP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20550-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
GPR100 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) and acts as a high affinity receptor for INSL5. Also acts as receptor for RLN3/relaxin-3, as well as bradykinin and kallidin. Binding of the ligand inhibits cAMP accumulation.Spezifikation
RXFP4 | |
Polyclonal | |
Unconjugated | |
RXFP4 | |
G protein coupled receptor 100, GPCR142, GPR100, Relaxin 3 receptor 2, RLN3 receptor 2, RLN3R2, RXFP4, RXFPR4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
339403 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TDU9 | |
RXFP4 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Liquid |