missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RRAGC Recombinant Protein
Marke: Abnova™ H00064121-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH16668 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
69.63 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MSLQYGAEETPLAGSYGAADSFPKDFGYGVEEEEEEAAAAGGGVGAGAGGGCGPGGADSSKPRILLMGLRRSGKSSIQKVVFHKMSPNETLFLESTNKIYKDDISNSSFVNFQIWDFPGQMDFFDPTFDYEMIFRGTGALIYVIDAQDDYMEALTRLHITVSKAYKVNPDMNFEVFIHKVDGLSDDHKIETQRDIHQRANDDLADAGLEKLHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSSPVDMQSYELCCDMIDVVIDVSCIYGLKEDGSGSAYDKESMAIIKLNNTTVLYLKEVTKFLALVCILREESFERKGLIDYNFHCFRKAIHEVFEVGVTSHRSCGHQTSASSLKALTHNGTPRNAI | |
FLJ13311/GTR2/RAGC | |
RRAGC | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
64121 | |
RRAGC (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RRAGC | |
Human | |
Recombinant | |
Solution |