missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11271-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL13 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13E family of ribosomal proteins. It is located in the cytoplasm. This gene is expressed at significantly higher levels in benign breast lesions than in breast carcinomas. Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13E family of ribosomal proteins. It is located in the cytoplasm. This gene is expressed at significantly higher levels in benign breast lesions than in breast carcinomas. Transcript variants derived from alternative splicing and/or alternative polyadenylation exist; these variants encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Spezifikation
RPL13 | |
Polyclonal | |
Unconjugated | |
RPL13 | |
60S ribosomal protein L13, BBC1, D16S444E, ribosomal protein L13, RPL13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
270106, 6137, 81765 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P26373, P41123, P47963 | |
Rpl13 | |
RPL13 Fusion Protein Ag1814 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |