missing translation for 'onlineSavingsMsg'
Learn More

RPH3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen RPH3A
Konzentration 0.2 mg/mL
Anwendungen Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16848443
Unterlagen ansehen Aktionsdetails
Proteintech
11396-1-AP-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 02-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16838443
Unterlagen ansehen Aktionsdetails
Proteintech
11396-1-AP-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 02-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The carboxy-terminal of rabphilin-3A consists of two C2 domains, A and B,and the amino-terminal (residues 45-170) contains a cysteine-rich rich region with two zinc finger motifs. Rabphilin-3A belongs to a family of other carboxyterminal type (C-type) tandem C2 proteins, which includes synaptotagmins and Doc2. Rabphilin is expressed in neuroendocrine cells and co-localizes with Rab3A on synaptic vesicles and chromaffin granules. Rabphilin-3A binds Rab3a/GTP/Mg+ within amino-terminal residues 45 and 170. Rabphilin-3A binds calcium ions and phosphatidylinositol 4,5-bisphophate containing lipid vesicles within its C2 domains. Rabphilin-3A is a positive regulator of calcium dependent exocytosis, while Rab3a is a negative regulator of exocytosis. Although rabphilin-3A associates with Rab3a, they seem to influence exocytosis independently of each other. Rabphilin-3A effects are likely mediated through interactions with an unknown factor that recognizes the Rab3 binding domain.
Spezifikation

Spezifikation

RPH3A
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
P47708, P47709, Q9Y2J0
171039, 19894, 22895
RPH3A Fusion Protein Ag1956
Primary
-20°C
RPH3A
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Exophilin 1, KIAA0985, Rabphilin 3A, rabphilin 3A homolog (mouse), RPH3A
Rph3a
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications