Learn More
Gilac™ Round Selective Sorting Dustbin
Round Selective Sorting Dustbin is great as a highly durable recycling bin. Reinforced bottom with anti-wear skirt.
Marke: Gilac™ G612021-1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
- Round dustbin great as a recycling bin.
- Highly durable.
- Reinforced bottom with anti-wear skirt.
- Reinforced handles to prevent distortion during carrying.
- Optional, choice of two lids.
- External Diameter includes handles.
- Width corresponds to diameter without handles.
Specifications
75 L | |
26.4 in. | |
White | |
Rund | |
19.1 in. | |
Dust Bin |
670 mm | |
19.8 gal. | |
485 mm | |
Nicht autoklavierbar | |
HDPE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gilac™ Round Selective Sorting Dustbin > Capacity: 75 L, Color: White
Spot an opportunity for improvement?Share a Content Correction