Learn More
STEMCELL Technologies RosetteSep™ Human Multiple Myeloma Cell Enrichment Cocktail
Artikelnummer. 17117681
Encompass
Ansicht ändern
Click to view available options
Menge:
200 ml Knochenmark
40 mL Knochenmark
Packungsgröße:
1 Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17117681 40 mL Knochenmark 1 Set
17177681 200 ml Knochenmark 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17117681 Lieferant STEMCELL Technologies Lieferanten-Nr. 15129
Please to purchase this item. Need a web account? Register with us today!
Immunodensity isolation of untouched plasma cells for multiple myeloma research
  • The RosetteSep™ Human Multiple Myeloma Cell Enrichment Cocktail is designed to enrich multiple myeloma cells (B cell and plasma cells) from fresh bone marrow aspirates by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD14, CD33, CD41, CD45RA, CD66b and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The purified multiple myeloma cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium
Spezifikation
Zelltyp B Cells, Plasma
Zur Verwendung mit (Anwendung) Cell Isolation
Menge 40 mL Knochenmark