missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | RNASE13 |
---|---|
Konzentration | 0.21 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16838464
|
Proteintech
18408-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16828464
|
Proteintech
18408-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Ribonucleases are ubiquitous enzymes involved in RNA metabolism and are classified in several families on the basis of their structural, catalytic and biological properties. RNase 13 (ribonuclease, RNase A family, 13), is a 156 amino acid secreted protein encoded by a gene that maps to human chromosome 14q11.2. Chromosome 14 houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer′s disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.Spezifikation
RNASE13 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q5GAL7, Q5GAM7, Q5GAN3 | |
440163, 497071, 497194 | |
RNASE13 Fusion Protein Ag12320 | |
Primary | |
-20°C | |
RNASE13 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Ribonuclease like protein 13, RNASE13 | |
RNASE13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |