missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RMP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11277-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The function of the C19orf2 protein remains unknown.The protein encoded by this gene binds to RNA polymerase II subunit 5 (RPB5) and negatively modulates transcription through its binding to RPB5. The encoded protein seems to have inhibitory effects on various types of activated transcription, but it requires the RPB5-binding region. This protein acts as a corepressor. It is suggested that it may require signaling processes for its function or that it negatively modulates genes in the chromatin structure. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Spezifikation
RMP | |
Polyclonal | |
Unconjugated | |
URI1 | |
C19orf2, NNX3, PPP1R19, Protein NNX3, RMP, RPB5 mediating protein, URI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19777, 8725 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94763, Q3TLD5 | |
URI1 | |
RMP Fusion Protein Ag1789 | |
150 μL | |
Primary | |
Human, Mouse | |
Liquid |