Learn More
MRPL38 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554704
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FHGATFVPRV PLHVAYAVGE DDLMPVYCGN EVTPTEAAQA PEVTYEAEEG SLWTLLLTSL DGHLLEPDAE YLHWLLTNIP GNRVAE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
MRPL38 Polyclonal antibody specifically detects MRPL38 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
MRPL38 | |
Polyclonal | |
Unconjugated | |
MRPL38 | |
1110036N21Rik; 39S ribosomal protein L38, mitochondrial; 4733401F03Rik; HSPC262; L38mt; Mitochondrial large ribosomal subunit protein mL38; mitochondrial ribosomal protein L38; MRP-L3; MRPL38; MRP-L38; R75240; ribosomal protein, mitochondrial, L3; rm38; Rpml3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64978 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DV4 | |
MRPL38 | |
Recombinant protein corresponding to Human RM38 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only