Learn More
Brady™ Rigid Polypropylene Class AB Fire Extinguisher Pictogram Sign
Product Code. 11760324 Shop All Brady Products
Change view
Click to view available options
Abmessungen (L x B):
200 x 250 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Abmessungen (L x B)
11760324 200 x 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11760324 Supplier Brady™ Supplier No. 252084
  • ONLINE EXCLUSIVE PRICE
    9.85 CHF
    8.78 CHF / 1 Stück
    Sparen 1.07 CHF
    10% Rabatt
Verfügbar ab: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Alternate Part Number STF 596-200X250-B7527
Höhe (metrisch) 250 mm
Länge (metrisch) 200 mm
Abmessungen (L x B) 200 x 250 mm