missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11717-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during maligt cellular transformation. Blood group related antigens represent a group of carbohydrate determits carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.Spezifikation
RHOD | |
Polyclonal | |
Unconjugated | |
RHOD | |
ARHD, Rho, Rho related protein HP1, RHOD, RHOHP1, RHOM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29984 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00212 | |
RHOD | |
RHOD Fusion Protein Ag2291 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |