Learn More
RHOBTB3 Mouse anti-Human, Unconjugated, Clone: 1H2E3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 67502-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The RHOBTB3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RHOBTB3. This antibody recognizes human, pig, mouse antigen. The RHOBTB3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
| RHOBTB3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1H2E3 | |
| Unconjugated | |
| 150 μL | |
| 22836 | |
| IgG1 |