missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhesus IL-18 Recombinant Protein

Artikelnummer. 11888811
Click to view available options
Menge:
20 μg
Packungsgröße:
20 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11888811

Marke: Invitrogen™ RRMIL18I

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Mature rhesus macaque IL-18 was expressed in Pichia pastoris, X-33, GenBank Accession # AAK13416, amino acids 37 - 193. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: YFGKLESKLS. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant rhesus macaque IL-18 was determined by its ability to induce IFNg production from KG-1 cells in the presence of recombinant mouse IL-12. 270 pg/mL and 470 pg/mL of IFNg are induced by 20 ng/mL and 40 ng/mL of recombinant rhesus macaque IL-18, respectively. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Spezifikation

Zur Verwendung mit (Anwendung) Control, ELISA, Immunoprecipitation, Western Blot
Zusammensetzung PBS with no preservative
Gen-ID (Entrez) 574151
Molekulargewicht 18.3 kDa
Name Rhesus IL-18
Menge 20 μg
Quelle Yeast
Kennzeichnung RUO
Gen-Alias cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1 g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320
Gebräuchliche Bezeichnung IL-18
Gensymbol IL18
Biologische Aktivität ED50 = 20 - 40 ng/mL; determined by the dose-dependent induction of IFN-gamma production from KG-1 cells in the presence or recombinant mouse IL-12.
Konjugat Unconjugated
Spezies Yeast
Rekombinant Recombinant
Sequenz Rhesus macaque IL-18, amino acids 37-193 (Accession # AAK13416)
Inhalt und Lagerung 4°C
Expressionssystem P. pastoris
Form Lyophilized
Protein-Subtyp Recombinant
Reinheits- oder Qualitätsgrad >95% by SDS-PAGE
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt