Learn More
RHBG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543367
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LATHEAYGDG LESVFPLIAE GQRSATSQAM HQLFGLFVTL MFASVGGGLG Sequence homology: Cow: 100%; Dog: 79%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%
RHBG Polyclonal antibody specifically detects RHBG in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
RHBG | |
Polyclonal | |
Unconjugated | |
RHBG | |
Ammonium transporter Rh type B; Rh family B glycoprotein; Rh family B glycoprotein (gene/pseudogene); rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhbg; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein; Rhesus blood group-associated B glycoprotein; SLC42A2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57127 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H310 | |
RHBG | |
Synthetic peptide directed towards the C-terminal of human RHBG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |