missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11904-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a putative retinal G-protein coupled receptor. The gene is a member of the opsin subfamily of the 7 transmembrane, G-protein coupled receptor 1 family. Like other opsins which bind retinaldehyde, it contains a conserved lysine residue in the seventh transmembrane domain. The protein acts as a photoisomerase to catalyze the conversion of all-trans-retinal to 11-cis-retinal. The reverse isomerization occurs with rhodopsin in retinal photoreceptor cells. The protein is exclusively expressed in tissue adjacent to retinal photoreceptor cells, the retinal pigment epithelium and Mueller cells. This gene may be associated with autosomal recessive and autosomal domit retinitis pigmentosa (arRP and adRP, respectively). Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
RGR | |
Polyclonal | |
Unconjugated | |
RGR | |
RGR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
306307, 57811, 5995 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P47804, Q9Z2B3 | |
Rgr | |
RGR Fusion Protein Ag2552 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |