missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGMA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12387-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Member of the repulsive guidance molecule (RGM) family that performs several functions in the developing and adult nervous system. Regulates cephalic neural tube closure, inhibits neurite outgrowth and cortical neuron branching, and the formation of mature synapses. Binding to its receptor NEO1/neogenin induces activation of RHOA-ROCK1/Rho-kinase signaling pathway through UNC5B-ARHGEF12/LARG-PTK2/FAK1 cascade, leading to collapse of the neuronal growth cone and neurite outgrowth inhibition. Furthermore, RGMA binding to NEO1/neogenin leads to HRAS inactivation by influencing HRAS-PTK2/FAK1-AKT1 pathway. It also functions as a bone morphogenetic protein (BMP) coreceptor that may signal through SMAD1, SMAD5, and SMAD8.Spezifikation
RGMA | |
Polyclonal | |
Unconjugated | |
RGMA | |
Repulsive guidance molecule A, RGM, RGM domain family member A, RGM domain family, member A, RGMA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
244058, 308739, 56963 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PCX7, Q96B86 | |
RGMA | |
RGMA Fusion Protein Ag3058 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |