missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REXO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18890-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
REXO4 | |
Polyclonal | |
Unconjugated | |
REXO4 | |
Exonuclease XPMC2, hPMC2, PMC2, REX4, REXO4, RNA exonuclease 4, XPMC2, XPMC2H | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227656, 311826, 57109 | |
Antibody | |
IgG |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6PAQ4, Q9GZR2 | |
Rexo4 | |
REXO4 Fusion Protein Ag13473 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Liquid |