Learn More
Thermo Scientific™ Cryptosporidium Controls
Artikelnummer. 12739260
Ansicht ändern
Click to view available options
Menge:
2 Fläschchen/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
12739260 2 Fläschchen/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 12739260 Lieferant Thermo Scientific™ Lieferanten-Nr. R2451030
  • ONLINE EXCLUSIVE PRICE
    111.00 CHF
    92.18 CHF / 1 Stück
    Sparen 18.82 CHF
    16% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 30-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Giardia/Cryptosporidium control test kits with Thermo Scientific Xpect Remel Cryptosporidium Controls.
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Giardia/Cryptosporidium control test kits with Thermo Scientific Xpect Remel Cryptosporidium Controls These kits contain positive and negative controls and are intended for QC purposes, to confirm that the test has been performed satisfactorily and can produce the expected result.
Cryptosporidium is a protozoan pathogen of the Apicomplexa phylum and causes a diarrheal illness called cryptosporidiosis. Cryptosporidiosis is typically an acute short-term infection but can become severe and non-resolving in children and immunocompromised individuals such as AIDS patients.
This kit contains Cryptosporidium positive control which is derived from 10% Formalin or SAF fixative stool samples of human or animal origin which are positive for Cryptosporidium; Giardia positive control derived from 10% formalin or SAF fixed stool samples which are positive for Giardia and Giardia/Cryptosporidium negative control which is derived from 10% Formalin or SAF fixative stool samples, which are negative for Giardia and Cryptosporidium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Spezifikation
Beschreibung Xpect™ Cryptosporidium Control Kit
Enthält 1 positive vial and 1 negative vial
Menge 2 Fläschchen/Kit