Learn More
Bellingham and Stanley™ Refractometer 0 to 30% starch Bellingham and Stanley
The Eclipse refractometer is a low cost, high quality hand-held refractometer robustly constructed for every day use.
Marke: Bellingham and Stanley™ 45-26
Additional Details : Gewicht : 0.01000kg
Beschreibung
The Eclipse refractometer is a low cost, high quality hand-held refractometer robustly constructed for every day use.
Spezifikation
Handheld Digital Refractometer | |
0 to 30 Brix |