missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recoverin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3B8E8, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66521-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Seems to be implicated in the pathway from retinal rod guanylate cyclase to rhodopsin. May be involved in the inhibition of the phosphorylation of rhodopsin in a calcium-dependent manner. The calcium-bound recoverin prolongs the photoresponse.Spezifikation
Recoverin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P34057, P35243 | |
RCVRN | |
Recoverin Fusion Protein Ag0119 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Liquid |
Western Blot | |
3B8E8 | |
Unconjugated | |
RCVRN | |
Protein CAR, RCV1, RCVRN, recoverin | |
Mouse | |
Protein G | |
RUO | |
100516153, 140936, 19674, 5957 | |
Antibody | |
IgG1 |