missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus VEGF E (Orf Virus) Protein
A cDNA sequence encoding the VEGF E (Orf Virus) was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7380.00 CHF
Spezifikation
Name | Virus VEGF E (Orf Virus) Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | The biological activity was determined (1) by the ability to induce VEGFR-2/KDR receptor phosphorylation in PAE/KDR cells and (2) in a cell proliferation assay using primary HUVECs. The ED50 for this effect is typically 1-5ng/ml. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15985548
|
enQuireBio™
QP10912-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965548
|
enQuireBio™
QP10912-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975548
|
enQuireBio™
QP10912-1MG |
1 mg |
7380.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
Virus VEGF E (Orf Virus) Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKLLVGILVA VCLHQYLLNA DSNTKGWSEV LKGSECKPRP IVVPVSETHP ELTSQRFNPP CVTLMRCGGC CNDESLECVP TEEVNVTMEL LGASGSGSNG MQRLSFVEHK KCDCRPRFTT TPPTTTRPPR RRR | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
The biological activity was determined (1) by the ability to induce VEGFR-2/KDR receptor phosphorylation in PAE/KDR cells and (2) in a cell proliferation assay using primary HUVECs. The ED50 for this effect is typically 1-5ng/ml. | |
Virus | |
His | |
The protein was lyophilized from a concentrated (1 mg/ml) solution containing PBS. |