missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Virus Norovirus Group-2 Protein

A cDNA sequence encoding the Norovirus Group-2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12867-100ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15991789

  • 227.00 CHF / 100 Mikrogramm
Verfügbar ab: 02-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

Virus Norovirus Group-2 Protein
Research Use Only
Recombinant Protein
E. coli
MKMASNDATP SNDGAAGLVP EINNEAMALD PVAGAAIAAP LTGQQNIIDP WIMNNFVQAP GGEFTVSPRN SPGEVLLNLE LGPEINPYLA HLARMYNGYA GGFEVQVVLA GNAFTAGKII FAAIPPNFPI ENLSAAQITM CPHVIVDVRQ LEPVNLPMPD VRNNFFHYNQ GSDSKLRLVA MLYTPLRANN SGDDVFTVSC RVLTRPSPDF SFNFLVPPTV ESKTKPFTLP ILTISEMSNS RFPVPIDSLH TSPTENIVVQ CQNGRVTLDG ELMGTTQLLP SQICAFRGVL TRSTSRASDQ ADTATPRLFN YYWHIQLDNL NGTPYDPAED IPGPLGTPDF RGKVFGVASQ RNPDSTTRAH EAKVDTTAGR FTPKLGSLEI STESGDFDQN QPTRFTPVGI GVDHEADFQQ WSLPDYSGQF THNMNLAPAV APNFPGEQLL FFRSQLPSSG GRSNGILDCR VPQEWVQHFY QESAPAQTQV ALVRYVNPDT GRVLFEAKLH KLGFMTIAKN GDSPITVPPN GYFRFESWVN PFYTLAPMGT GNGRRRIQ
Protein is >85% pure as determined by 12% PAGE (coomassie staining).
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Virus
His
Phosphate buffer, 1M urea and 50mM arginine.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote