missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant SARS SARS Envelope Protein
A cDNA sequence encoding the SARS Envelope was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13417-500ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
SARS SARS Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 60mM NaCl, and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
SARS | |
Untagged | |
SARS Envelope protein is >95% pure as determined by 10% PAGE(coomassie staining). |