missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rubella Virus Rubella E1 Protein
A cDNA sequence encoding the Rubella E1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13373-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Rubella Virus Rubella E1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1xpbs. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
ubella Virus | |
GST | |
Protein is >90% pure as determined by SDS-PAGE. |