missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rubella Virus Rubella Capsid Protein
A cDNA sequence encoding the Rubella Capsid was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13372-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Rubella Virus Rubella Capsid Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 10mM glutation, 0.2% sarcosyl and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
ubella Virus | |
GST | |
Rubella protein is >95% pure as determined by 10% PAGE (coomassie staining). |