missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque S100b Protein
A cDNA sequence encoding the S100b was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13401-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
S100b Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SELEKAMVAL IDVFHQYSGR EGDKHKLKKS ELKELINNEL SHFLEEIKEQ EVVDKVMETL DSDGDGECDF QEFMAFVAMV TTACHEFFEH E | |
Greater than 97.0% as determined by SDS-PAGE and HPLC analyses. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2um filtered concentrated solution in PBS, pH 7.4. |