missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque Rantes Protein
A cDNA sequence encoding the Rantes was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Gen-ID (Entrez) | 574178 |
---|---|
Name | Rantes Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gensymbol | CCL5 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15975378
|
enQuireBio™
QP10857-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965378
|
enQuireBio™
QP10857-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15955378
|
enQuireBio™
QP10857-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
574178 | |
Research Use Only | |
CCL5 | |
Recombinant Protein | |
E. coli | |
SPHASDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Rantes Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 1.0-10 ng/ml. | |
Rhesus Macaque | |
Untagged | |
Rantes protein was lyophilized from a 0.2microm filtered concentrated solution in 20mM PB pH 6.0 and 500mM NaCl. |