missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SCGN Protein
A cDNA sequence encoding the SCGN was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13431-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
SCGN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS MDNAHRQTQA HLDAACFWQI WQRFDKDEKG YIKETELDAF FDDLLAKFGI EDTLMEENVQ KMKEQLMVGH DISKEGRILM KELASMFLSE DENFLLFFRL ETPLDNSVEF MQIWRKYDAD SSGFISAAEL SNFLRDLFLH HKKVISEAEL EEYTSTMMKI FDRNKDGRLD LNDLARILAL QENFLLQFKM DASSTEERKR DFEKIFAHYD VSKTGALEGP EVDGFVKDMM ELVQPSISGV DLDKFREILL RHCDVNKDGK IQKSELALCLGLKINP | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
The sterile filtered concentrated protein solution was lyophilized with 20mM Tris and 50mM NaCl pH 7.5. |