missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat NME2 Protein
A cDNA sequence encoding the NME2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12854-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NME2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMANLERT FIAIKPDGVQ RGLVGEIIKR FEQKGFRLVA MKFLRASEEH LKQHYIDLKD RPFFPGLVKY MNSGPVVAMV WEGLNVVKTG RVMLGETNPA DSKPGTIRGD FCIQVGRNII HGSDSVESAE KEIGLWFKPE ELIDYKSCAH DWVYE | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
NME2 protein solution (1 mg/ml) containing Phosphate Buffered Saline pH 7.4 and 10% glycerol. |