missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat LDHA Protein
A cDNA sequence encoding the LDHA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12549-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
24533 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MAALKDQLIV NLLKEEQVPQ NKITVVGVGA VGMACAISIL MKDLADELAL VDVIEDKLKG EMMDLQHGSL FLKTPKIVSS KDYSVTANSK LVIITAGARQ QEGESRLNLV QRNVNIFKFI IPNVVKYSPQ CKLLIVSNPV DILTYVAWKI SGFPKNRVIG SGCNLDSARF RYLMGERLGV HPLSCHGWVL GEHGDSSVPV WSGVNVAGVS LKSLNPQLGT DADKEQWKDV HKQVVDSAYE VIKLKGYTSW AIGLSVADLA ESIMKNLRRV HPISTMIKGL YGIKEDVFLS VPCILGQNGI SDVVKVTLTP DEEARLKKSA DTLWGIQKEL QFLEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
LDHA Protein | |
Research Use Only | |
Ldha | |
Rat | |
His | |
LDHA protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 1mM DTT and 20% glycerol. |