missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 33 Protein
A cDNA sequence encoding the IL 33 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12399-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
361749 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSIQGTSL LTESCALSTY NDQSVSFVLE NGCYVINVED CGKNQEKDKV LLRYYESSFP AQSGDGVDGK KLMVNMSPIK DTDIWLNAND KDYSVELQKG DVSPPDQAFF VLHKKSSDFV SFECKNLPGT YIGVKDNQLA LVEENDESCN NIMFKLSKM | |
Greater than 90.0% as determined by SDS-PAGE. |
IL 33 Protein | |
Research Use Only | |
Il33 | |
Rat | |
His | |
IL 33 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |