missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 21 Protein
A cDNA sequence encoding the IL 21 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10702-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
365769 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity is measured by its ability to proliferate activated B cells. | |
Rat | |
Untagged | |
Rat IL-21 was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
IL 21 Protein | |
Research Use Only | |
Il21 | |
Recombinant Protein | |
E. coli | |
HKSSPQRPDH LLIRLRHLMD IVEQLKIYEN DLDPELLTAP QDVKGQCEHE AFACFQKAKL KPSNTGNNKT FINDLLAQLR RRLPAKRTGN KQRHMAKCPS CDLYEKKTPK EFLERLKWLL QKMIHQHLS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |