missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 1RA Protein
A cDNA sequence encoding the IL 1RA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12395-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
60582 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHPAGKRP CKMQAFRIWD TNQKTFYLRN NQLIAGYLQG PNTKLEEKID MVPIDFRNVF LGIHGGKLCL SCVKSGDDTK LQLEEVNITD LNKNKEEDKR FTFIRSETGP TTSFESLACP GWFLCTTLEA DHPVSLTNTP KEPCTVTKFY FQEDQ | |
Greater than 90.0% as determined by analysis by SDS-PAGE. |
IL 1RA Protein | |
Research Use Only | |
Il1rn | |
Rat | |
His | |
IL 1RA protein solution (1 mg/ml) containing Phosphate Buffer Saline (pH 7.4), 10% glycerol and 1mM DTT. |