missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL3 / GRO gamma Protein
A DNA sequence encoding the Rat CXCL3 / GRO gamma, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10282-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_612531.1 | |
CXCL3 / GRO gamma Protein | |
Research Use Only | |
CXCL3 | |
The biological activity determined by a chemotaxis bioassay using human CXCR2 transfected murine BaF3 cells is in a concentration range of 5-50 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
171551 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
RELRCQCLKT LPRVDFENIQ SLTVTPPGPH CTQTEVIATL KDGQEVCLNP QAPRLQKIIQ KLLKSDKSS | |
Greater than 96% as determined by SDS-PAGE and HPLC. |