missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL14 / BRAK Protein
A DNA sequence encoding the Rat CXCL14 / BRAK, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10289-ec-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001013155.1 | |
CXCL14 / BRAK Protein | |
Research Use Only | |
Cxcl14 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 1.0-10 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
306748 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIITTKS MSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE | |
Greater than 95% as determined by SDS-PAGE and HPLC. |