missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL1 / MGSA / NAP-3 Protein
A DNA sequence encoding the Rat CXCL1 / MGSA / NAP-3, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10280-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_110472.1 | |
CXCL1 / MGSA / NAP-3 Protein | |
Research Use Only | |
CXCL1 | |
The biological activity determined by a chemotaxis bioassay using rat neutrophils is in a concentration range of 10-100 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
81503 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APVANELRCQ CLQTVAGIHF KNIQSLKVMP PGPHCTQTEV IATLKNGREA CLDPEAPMVQ KIVQKMLKGV PK | |
Greater than 97% as determined by SDS-PAGE and HPLC. |