missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL3 / Mip1a Protein
A DNA sequence encoding the Rat CCL3 / Mip1a, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10292-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_037157.2 | |
CCL3 / Mip1a Protein | |
Research Use Only | |
CCL3 | |
The biological activity determined by a chemoattract bioassay using human blood monocytes is in a concentration range of 10-100 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
25542 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APYGADTPTA CCFSYGRQIP RKFIADYFET SSLCSQPGVI FLTKRNRQIC ADPKETWVQE YITELELNA | |
Greater than 98% as determined by SDS-PAGE and HPLC. |