missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cardiotrophin-1 / CTF1 Protein
A DNA sequence encoding the Rat Cardiotrophin-1 / CTF1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10167-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_058825.1 | |
Cardiotrophin-1 / CTF1 Protein | |
Research Use Only | |
Ctf1 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Rat | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
29201 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MSQREGSLED HQTDSSFSFL PHLEAKIRQT HNLARLLTKY ADQLLEEYVQ QQGEPFGLPG FSPPRLPLAG LSGPAPSHAG LPVSERLRQD AAALSALPAL LDAVRRRQAE LNPRAPRLLR SLEDAARQVR ALGAAVETVL AALGAAARGP VPEPVATSAL FTSNSAAGVF SAKVLGLHVC GLYGEWVSRT EGDLGQLVPG GVA | |
Greater than 95% as determined by SDS-PAGE and HPLC. |